2,3-Difluorisonicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluoropyridine-4-carboxylic acid

PC49549-5G PC49549-25G
APOSPC49549-5GEA 226 EUR
APOSPC49549-5G APOSPC49549-25G
2,3-Difluorisonicotinsäure 97%
2,3-Difluorisonicotinsäure
Formel: C₆H₃F₂NO₂
Schmelzpunkt: 163…168 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09031174
CAS-Nummer: 851386-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR