2,3-Difluorisonicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Difluoropyridine-4-carboxylic acid
PC49549-5G
PC49549-25G
APOSPC49549-5GEA
226
EUR
APOSPC49549-5G
APOSPC49549-25G
2,3-Difluorisonicotinsäure 97%
2,3-Difluorisonicotinsäure
Formel:
C₆H₃F₂NO₂ Schmelzpunkt: 163…168 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09031174 CAS-Nummer: 851386-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...