1,4-Dimethoxytetrafluorbenzol
Lieferant: Apollo Scientific
|
Achtung
|
PC4955-5G
APOSPC4955-5GEA
256
EUR
APOSPC4955-5G
1,4-Dimethoxytetrafluorbenzol
1,4-Dimethoxytetrafluorobenzene
Formel:
C₈H₆F₄O₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
362-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...