4-(2,2,2-Trifluoroethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-4-(2,2,2-trifluoroethyl)benzene, α-Bromo-4-(2,2,2-trifluoroethyl)toluene

PC49576-1G
APOSPC49576-1GEA 669 EUR
APOSPC49576-1G
4-(2,2,2-Trifluoroethyl)benzyl bromide 95%
4-(2,2,2-Trifluoroethyl)benzyl bromide
Formel: C₉H₈BrF₃
Molecular Weight: 253,059 g/mol
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140177
CAS-Nummer: 1221272-87-2
UN: 2928
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR