4-Bromo-3-(trifluoromethyl)benzene-1,2-diamine 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-2,3-diaminobenzotrifluoride, 4-Bromo-1,2-diamino-3-(trifluoromethyl)benzene
PC49609-250MG
APOSPC49609-250MGEA
174
EUR
APOSPC49609-250MG
4-Bromo-3-(trifluoromethyl)benzene-1,2-diamine 90%
4-Bromo-3-(trifluoromethyl)benzene-1,2-diamine
Formel:
C₇H₆BrF₃N₂ Molecular Weight: 255,035 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25541886 CAS-Nummer: 1445995-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...