2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC49614-1G
PC49614-250MG
APOSPC49614-1GEA
0
EUR
APOSPC49614-1G
APOSPC49614-250MG
2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid 97%
2-Hydroxy-5-iodo-3-(trifluoromethyl)benzoic acid
Formel:
C₈H₄F₃IO₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD25541889 CAS-Nummer: 1445995-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...