3-Fluoro-2-methoxyphenacyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC49619-1G PC49619-250MG
APOSPC49619-1GEA 0 EUR
APOSPC49619-1G APOSPC49619-250MG
3-Fluoro-2-methoxyphenacyl bromide 97%
3-Fluoro-2-methoxyphenacyl bromide
Formel: C₉H₈BrFO₂
Lagertemperatur: Kühlschrank
MDL: MFCD23711719
CAS-Nummer: 1427403-76-6
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR