5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%

Lieferant: Apollo Scientific
Gefahr

PC49623-1G PC49623-250MG
APOSPC49623-1GEA 0 EUR
APOSPC49623-1G APOSPC49623-250MG
5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile 97%
5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile
Formel: C₉H₅F₃INO
Lagertemperatur: Raumtemperatur
MDL: MFCD25541890
CAS-Nummer: 1445995-74-3
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR