1-(Difluoromethyl)-2,4-difluorobenzene

Lieferant: Apollo Scientific

PC49631-5G
APOSPC49631-5GEA 219 EUR
APOSPC49631-5G
1-(Difluoromethyl)-2,4-difluorobenzene
1-(Difluoromethyl)-2,4-difluorobenzene
Formel: C₇H₄F₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06657968
CAS-Nummer: 845866-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR