5-Brom-2-chlor-4-(trifluormethyl)pyridin

Lieferant: Apollo Scientific
Achtung

PC49632-1G
APOSPC49632-1GEA 40.6 EUR
APOSPC49632-1G
5-Brom-2-chlor-4-(trifluormethyl)pyridin
5-Brom-2-chlor-4-(trifluormethyl)pyridin
Formel: C₆H₂BrClF₃N
Molecular Weight: 260,44 g/mol
Siedepunkt: 86…88 °C/19mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD11100520
CAS-Nummer: 823221-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR