4-Chloro-2-(trifluoromethyl)nicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

PC49634-250MG PC49634-1G PC49634-5G
APOSPC49634-250MGEA 144 EUR
APOSPC49634-250MG APOSPC49634-1G APOSPC49634-5G
4-Chloro-2-(trifluoromethyl)nicotinic acid 95%
4-Chloro-2-(trifluoromethyl)nicotinic acid
Formel: C₇H₃ClF₃NO₂
MDL: MFCD22546902
CAS-Nummer: 1018678-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR