2,6-Difluoro-3-(trifluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC49654-1G
APOSPC49654-1GEA
205
EUR
APOSPC49654-1G
2,6-Difluoro-3-(trifluoromethyl)pyridine 97%
2,6-Difluoro-3-(trifluoromethyl)pyridine
Formel:
C₆H₂F₅N Siedepunkt: 70…71 °C/100mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546949 CAS-Nummer: 58584-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...