Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%

Lieferant: Apollo Scientific

PC49657-1G
APOSPC49657-1GEA 56.7 EUR
APOSPC49657-1G
Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Methyl 4-bromo-2-chloro-6-fluorobenzoate
Formel: C₈H₅BrClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22574196
CAS-Nummer: 1321613-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR