2-(Aminomethyl)-6-fluoroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Fluoroimidazo[1,2-a]pyridin-2-yl)methylamine

PC49658-250MG
APOSPC49658-250MGEA 358 EUR
APOSPC49658-250MG
2-(Aminomethyl)-6-fluoroimidazo[1,2-a]pyridine
2-(Aminomethyl)-6-fluoroimidazo[1,2-a]pyridine
Formel: C₈H₈FN₃
Molecular Weight: 165,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09994905
CAS-Nummer: 1020033-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR