2-Bromo-5-chloro-4-fluoronitrobenzene
Lieferant: Apollo Scientific
Synonyme:
1-Bromo-4-chloro-5-fluoro-2-nitrobenzene
PC49659-5G
APOSPC49659-5GEA
92.8
EUR
APOSPC49659-5G
2-Bromo-5-chloro-4-fluoronitrobenzene
2-Bromo-5-chloro-4-fluoronitrobenzene
Formel:
C₆H₂BrClFNO₂ Molecular Weight: 254.441 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18089309 CAS-Nummer: 960000-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...