2-Chloro-6-(trifluoromethyl)isonicotinonitrile

Lieferant: Apollo Scientific
Achtung

PC49663-1G
APOSPC49663-1GEA 157 EUR
APOSPC49663-1G
2-Chloro-6-(trifluoromethyl)isonicotinonitrile
2-Chloro-6-(trifluoromethyl)isonicotinonitrile
Formel: C₇H₂ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13190042
CAS-Nummer: 1196155-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR