Methyl 2,3-difluoro-5-methylbenzoate

Lieferant: Apollo Scientific
Achtung

PC49668-1G PC49668-250MG
APOSPC49668-1GEA 575 EUR
APOSPC49668-1G APOSPC49668-250MG
Methyl 2,3-difluoro-5-methylbenzoate
Methyl 2,3-difluoro-5-methylbenzoate
Formel: C₉H₈F₂O₂
Molecular Weight: 186.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042838
CAS-Nummer: 1314919-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR