(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-trans-D-4-F-Pro-OH, (2R,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid
PC49679-250MG
APOSPC49679-250MGEA
27.7
EUR
APOSPC49679-250MG
(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected 95+%
(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid, N-BOC protected
Formel:
C₁₀H₁₆FNO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08706411 CAS-Nummer: 681128-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...