2-Bromo-6-(difluoromethyl)benzonitrile 95%

Lieferant: Apollo Scientific
Gefahr

PC49699-1G
APOSPC49699-1GEA 522 EUR
APOSPC49699-1G
2-Bromo-6-(difluoromethyl)benzonitrile 95%
2-Bromo-6-(difluoromethyl)benzonitrile
Formel: C₈H₄BrF₂N
Molecular Weight: 232,02 g/mol
MDL: MFCD18391060
CAS-Nummer: 1261775-76-1
UN: 2811

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR