2,4,5-Trifluorbenzonitril 96%
Lieferant: Apollo Scientific
PC49703-100G
PC49703-25G
PC49703-5G
APOSPC49703-100GEA
156
EUR
APOSPC49703-100G
APOSPC49703-25G
APOSPC49703-5G
2,4,5-Trifluorbenzonitril 96%
2,4,5-Trifluorbenzonitril
Formel:
C₇H₂F₃N Molecular Weight: 157.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013289 CAS-Nummer: 98349-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...