2',3'-Difluoracetophenon 97%

Lieferant: Apollo Scientific

Synonyme: 1-(2,3-Difluorphenyl)ethan-1-on

PC49704-100G PC49704-25G PC49704-5G
APOSPC49704-100GEA 252 EUR
APOSPC49704-100G APOSPC49704-25G APOSPC49704-5G
2',3'-Difluoracetophenon 97%
2',3'-Difluoracetophenon
Formel: C₈H₆F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00061274
CAS-Nummer: 18355-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR