2-Bromo-4-(difluoromethyl)-1-iodobenzene 95%

Lieferant: Apollo Scientific

PC49705-1G PC49705-250MG
APOSPC49705-1GEA 0 EUR
APOSPC49705-1G APOSPC49705-250MG
2-Bromo-4-(difluoromethyl)-1-iodobenzene 95%
2-Bromo-4-(difluoromethyl)-1-iodobenzene
Formel: C₇H₄BrF₂I
Lagertemperatur: Raumtemperatur
MDL: MFCD18391106
CAS-Nummer: 1261642-33-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR