Ethyl 2-(5-amino-2-bromo-4-fluorophenyl)acetate 95%

Lieferant: Apollo Scientific

PC49719-250MG
APOSPC49719-250MGEA 522 EUR
APOSPC49719-250MG
Ethyl 2-(5-amino-2-bromo-4-fluorophenyl)acetate 95%
Ethyl 2-(5-amino-2-bromo-4-fluorophenyl)acetate
Formel: C₁₀H₁₁BrFNO₂
Molecular Weight: 276,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28405818
CAS-Nummer: 1442471-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR