4-Chlor-2-fluorphenylacetonitril 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-fluorobenzeneacetonitrile

PC49721-100G PC49721-25G PC49721-5G
APOSPC49721-100GEA 191 EUR
APOSPC49721-100G APOSPC49721-25G APOSPC49721-5G
4-Chlor-2-fluorphenylacetonitril 95%
4-Chlor-2-fluorphenylacetonitril
Formel: C₈H₅ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD01312758
CAS-Nummer: 75279-53-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR