2-Bromo-4-chloro-5-fluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC49811-1G PC49811-250MG
APOSPC49811-1GEA 638 EUR
APOSPC49811-1G APOSPC49811-250MG
2-Bromo-4-chloro-5-fluorobenzoic acid 95%
2-Bromo-4-chloro-5-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28733901
CAS-Nummer: 157165-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR