4-Fluor-3-(phenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC4984-1G
APOSPC4984-1GEA 205 EUR
APOSPC4984-1G
4-Fluor-3-(phenylcarbamoyl)phenylboronsäure 98%
4-Fluor-3-(phenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
Schmelzpunkt: 252…254 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235052
CAS-Nummer: 874219-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR