3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: (3-Fluoro-5-(piperidine-1-carbonyl)phenyl)boronic acid

PC4985-1G
APOSPC4985-1GEA 212 EUR
APOSPC4985-1G
3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure 98%
3-Fluor-5-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Molecular Weight: 251.0600 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235060
CAS-Nummer: 874219-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR