4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC4986-1G
APOSPC4986-1GEA 299 EUR
APOSPC4986-1G
4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure 98%
4-Fluor-3-(piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₅BFNO₃
Schmelzpunkt: 155…157 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235049
CAS-Nummer: 874219-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR