4-Fluor-3-(propylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-(N-propylcarbamoyl)phenylboronic acid
PC4988-1G
APOSPC4988-1GEA
205
EUR
APOSPC4988-1G
4-Fluor-3-(propylcarbamoyl)phenylboronsäure 98%
4-Fluor-3-(propylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₃BFNO₃ Schmelzpunkt: 105…107 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235051 CAS-Nummer: 874219-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...