3-Brom-5-(trifluormethoxy)phenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-3-hydroxy-5-(trifluormethoxy)benzol

PC499005-1G PC499005-250MG PC499005-5G
APOSPC499005-1GEA 162 EUR
APOSPC499005-1G APOSPC499005-250MG APOSPC499005-5G
3-Brom-5-(trifluormethoxy)phenol 97%
3-Brom-5-(trifluormethoxy)phenol
Formel: C₇H₄BrF₃O₂
Molecular Weight: 257.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142929
CAS-Nummer: 1197239-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR