3-Brom-5-(trifluormethoxy)phenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-hydroxy-5-(trifluormethoxy)benzol
PC499005-1G
PC499005-250MG
PC499005-5G
APOSPC499005-1GEA
162
EUR
APOSPC499005-1G
APOSPC499005-250MG
APOSPC499005-5G
3-Brom-5-(trifluormethoxy)phenol 97%
3-Brom-5-(trifluormethoxy)phenol
Formel:
C₇H₄BrF₃O₂ Molecular Weight: 257.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15142929 CAS-Nummer: 1197239-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...