3-Bromo-5-methoxybenzotrifluoride

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-3-methoxy-5-(trifluoromethyl)benzene, 1-Bromo-3-(trifluoromethyl)-5-methoxybenzene

PC499010-1G PC499010-25G PC499010-5G
APOSPC499010-1GEA 21.9 EUR
APOSPC499010-1G APOSPC499010-25G APOSPC499010-5G
3-Bromo-5-methoxybenzotrifluoride
3-Bromo-5-methoxybenzotrifluoride
Formel: C₈H₆BrF₃O
Molecular Weight: 255.032 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09999639
CAS-Nummer: 627527-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR