2-Chlor-3-fluorbenzaldehyd 98%
Lieferant: Apollo Scientific
PC499011-5G
APOSPC499011-5GEA
23.2
EUR
APOSPC499011-5G
2-Chlor-3-fluorbenzaldehyd 98%
2-Chloro-3-fluorobenzaldehyde
Formel:
C₇H₄ClFO Schmelzpunkt: 29…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782053 CAS-Nummer: 96516-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...