2-Chloro-3-fluorobenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-2-chloro-3-fluorobenzene
PC499012-1G
APOSPC499012-1GEA
21.9
EUR
APOSPC499012-1G
2-Chloro-3-fluorobenzyl bromide
2-Chloro-3-fluorobenzyl bromide
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol Schmelzpunkt: 41…44 °C Flammpunkt: 91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038462 CAS-Nummer: 874285-19-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...