4-Nitro-2-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific

PC499017-1G PC499017-250MG
APOSPC499017-1GEA 0 EUR
APOSPC499017-1G APOSPC499017-250MG
4-Nitro-2-(trifluoromethyl)phenylacetonitrile
4-Nitro-2-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13185782
CAS-Nummer: 1067192-61-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR