3-Fluor-5-(propylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-5-(propylcarbamoyl)phenylboronic acid

PC4994-1G
APOSPC4994-1GEA 205 EUR
APOSPC4994-1G
3-Fluor-5-(propylcarbamoyl)phenylboronsäure 98%
3-Fluor-5-(propylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Schmelzpunkt: 95…97 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235054
CAS-Nummer: 874219-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR