3-Benzylcarbamoyl-5-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Benzylcarbamoyl)-5-fluorophenylboronic acid

PC4995-1G
APOSPC4995-1GEA 205 EUR
APOSPC4995-1G
3-Benzylcarbamoyl-5-fluorphenylboronsäure 98%
3-Benzylcarbamoyl-5-fluorphenylboronsäure
Formel: C₁₄H₁₃BFNO₃
Schmelzpunkt: 263…265 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235058
CAS-Nummer: 874219-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR