3-(Cyclohexylcarbamoyl)-5-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

PC4996-1G
APOSPC4996-1GEA 205 EUR
APOSPC4996-1G
3-(Cyclohexylcarbamoyl)-5-fluorphenylboronsäure 98%
3-(Cyclohexylcarbamoyl)-5-fluorphenylboronsäure
Formel: C₁₃H₁₇BFNO₃
Schmelzpunkt: 239…241 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235059
CAS-Nummer: 874219-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR