3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
3-(Diethylcarbamoyl)-4-fluorophenylboronic acid, 5-Borono-N,N-diethyl-2-fluorobenzamide
PC4997-1G
APOSPC4997-1GEA
299
EUR
APOSPC4997-1G
3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid 98%
3-(Diethylcarbamoyl)-4-fluorobenzeneboronic acid
Formel:
C₁₁H₁₅BFNO₃ Molecular Weight: 239,051 g/mol Schmelzpunkt: 133…135 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235047 CAS-Nummer: 874219-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...