3-(Dimethylcarbamoyl)-5-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC4999-1G
APOSPC4999-1GEA 205 EUR
APOSPC4999-1G
3-(Dimethylcarbamoyl)-5-fluorphenylboronsäure 98%
3-(Dimethylcarbamoyl)-5-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Schmelzpunkt: 111…113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235056
CAS-Nummer: 874219-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR