6-Bromo-4,5-difluoro-1H-benzimidazole

Lieferant: Apollo Scientific

PC500001-1G
APOSPC500001-1GEA 677 EUR
APOSPC500001-1G
6-Bromo-4,5-difluoro-1H-benzimidazole
6-Bromo-4,5-difluoro-1H-benzimidazole
Formel: C₇H₃BrF₂N₂
Molecular Weight: 233,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369743
CAS-Nummer: 1375068-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR