2-Bromo-4-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC500002-1G PC500002-250MG PC500002-5G
APOSPC500002-1GEA 0 EUR
APOSPC500002-1G APOSPC500002-250MG APOSPC500002-5G
2-Bromo-4-(trifluoromethyl)benzyl bromide 97%
2-Bromo-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11847628
CAS-Nummer: 657-64-7
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR