2-Bromo-4-(trifluoromethyl)benzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC500002-1G
PC500002-250MG
PC500002-5G
APOSPC500002-1GEA
0
EUR
APOSPC500002-1G
APOSPC500002-250MG
APOSPC500002-5G
2-Bromo-4-(trifluoromethyl)benzyl bromide 97%
2-Bromo-4-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅Br₂F₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847628 CAS-Nummer: 657-64-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...