1-Brom-2,3-difluor-5-nitrobenzol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Brom-4,5-difluornitrobenzol

PC500003-1G
APOSPC500003-1GEA 37.7 EUR
APOSPC500003-1G
1-Brom-2,3-difluor-5-nitrobenzol
1-Bromo-2,3-difluoro-5-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Molecular Weight: 237.99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095339
CAS-Nummer: 374633-24-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR