4-(Trifluoromethyl)isatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
PC500015-1G
PC500015-250MG
PC500015-5G
APOSPC500015-1GEA
0
EUR
APOSPC500015-1G
APOSPC500015-250MG
APOSPC500015-5G
4-(Trifluoromethyl)isatoic anhydride
4-(Trifluoromethyl)isatoic anhydride
Formel:
C₉H₄F₃NO₃ Schmelzpunkt: 240…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD21946140 CAS-Nummer: 97928-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...