Trifluoroborane acetonitrile complex-Lösung
Lieferant: Apollo Scientific
|
Gefahr
|
PC50006-1KG
PC50006-250G
APOSPC50006-1KGEA
426
EUR
APOSPC50006-1KG
APOSPC50006-250G
Trifluoroborane acetonitrile complex-Lösung
Trifluoroborane acetonitrile complex
Formel:
BF₃·CH₃CN Dichte: 0,87 g/cm³ (25 °C) Flammpunkt: 10 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD28125563 CAS-Nummer: 420-16-6 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...