2-Nitro-4-(trifluoromethyl)benzyl chloride 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5001-1G
APOSPC5001-1GEA
87.3
EUR
APOSPC5001-1G
2-Nitro-4-(trifluoromethyl)benzyl chloride 98%
2-Nitro-4-(trifluoromethyl)benzyl chloride
Formel:
C₈H₅ClF₃NO₂ Schmelzpunkt: 42…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631694 CAS-Nummer: 225656-59-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...