(3,4-Difluorophenyl)methanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC50010-250MG PC50010-1G
APOSPC50010-250MGEA 0 EUR
APOSPC50010-250MG APOSPC50010-1G
(3,4-Difluorophenyl)methanesulphonyl chloride
(3,4-Difluorophenyl)methanesulphonyl chloride
Formel: C₇H₅ClF₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08704570
CAS-Nummer: 163295-73-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR