(3,4-Difluorophenyl)methanesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC50010-250MG
PC50010-1G
APOSPC50010-250MGEA
0
EUR
APOSPC50010-250MG
APOSPC50010-1G
(3,4-Difluorophenyl)methanesulphonyl chloride
(3,4-Difluorophenyl)methanesulphonyl chloride
Formel:
C₇H₅ClF₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704570 CAS-Nummer: 163295-73-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...