(3,5-Difluorphenyl)methansulfonsäurefluorid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,5-Difluoro-α-toluenesulphonyl chloride

PC50011-1G PC50011-250MG
APOSPC50011-1GEA 87 EUR
APOSPC50011-1G APOSPC50011-250MG
(3,5-Difluorphenyl)methansulfonsäurefluorid 97%
(3,5-Difluorphenyl)methansulfonsäurefluorid
Formel: C₇H₅ClF₂O₂S
Molecular Weight: 226.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743349
CAS-Nummer: 163295-74-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR